missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBBP1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38094-100ul
This item is not returnable.
View return policy
Description
MYBBP1A Polyclonal antibody specifically detects MYBBP1A in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| MYBBP1A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| MYB binding protein (P160) 1a, myb-binding protein 1A, P160FLJ37886, p53-activated protein-2, PAP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1104-1328 of human MYBBP1A (NP_055335.2).,, Sequence:, DLTVLLGVLQGQQQSLQQGAHSTGSSRLHDLYWQAMKTLGVQRPKLEKKDAKEIPSATQSPISKKRKKKGFLPETKKRKKRKSEDGTPAEDGTPAATGGSQPPSMGRKKRNRTKAKVPAQANGTPTTKSPAPGAPTRSPSTPAKSPKLQKKNQKPSQVNGAPGSPTEPAGQKQHQKALPKKGVLGKSPLSALARKKARLSLVIRSPSLLQSGAKKKAQVRKAGKP | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10514 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction