missing translation for 'onlineSavingsMsg'
Learn More

MXD4 Monoclonal Antibody (OTI7F7), TrueMAB™, OriGene

Product Code. 17500704
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17500704 30 μL 30µL
17580694 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17500704 Supplier Origene Technologies Supplier No. TA808928S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MXD4 Monoclonal antibody specifically detects MXD4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MXD4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HDGF
Gene Accession No. P51858
Gene Alias 2810410A03Rik; bHLHc12; Class C basic helix-loop-helix protein 12; LOW QUALITY PROTEIN: max dimerization protein 4; MAD4; Mad4 homolog; max dimerization protein 4; Max dimerizer 4; max-associated protein 4; Max-interacting transcriptional repressor MAD4; MST149; MSTP149; MXD4
Host Species Mouse
Immunogen Full length human recombinant protein of human HDGF produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3068
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.