missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MxA/Mx1 Antibody (MX1/7529), Novus Biologicals™

Mouse Monoclonal Antibody

245.00€ - 486.00€

Specifications

Antigen MxA/Mx1
Clone MX1/7529
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043033
View Documents
Novus Biologicals™
NBP3-26822-20ug
20 μg
245.00€
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043805
View Documents
Novus Biologicals™
NBP3-26822-100ug
100 μg
486.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MxA/Mx1 Monoclonal antibody specifically detects MxA/Mx1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MxA/Mx1
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis
10 mM PBS with 0.05% BSA
4599
IgG2 κ
Protein A or G purified
MX1/7529
Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Monoclonal
Purified
RUO
Human
human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse)
Recombinant fragment (around aa400-592) of human MxA/Mx1 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.