missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUTYH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | MUTYH |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16899124
|
Proteintech
19650-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16889124
|
Proteintech
19650-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYH (mutY homolog (E. coli )) is a DNA glycosylase mismatch repair enzyme that in conjunction with mutM (OGG1), cleaves adenine residues paired with either oxidized (8-hydroxyguanines) or non-modified guanines in order to correct A/G and A/C mismatches. Repair of most modified and mispaired bases in the genome is initiated by DNA glycosylases, which bind and cleave N-glycosyl bonds to initiate base excision repair. MYH is crucial for the avoidance of mutations resulting from oxidative DNA damage. Multiple N-terminal splice variants of MYH exist in mammalian cells. Increasing levels of MYH in A549 cells exposed to oxygen and infrared radiation leads to improvements in cell survival. Biallelic MYH germ-line mutations predispose humans to colorectal adenomas and carcinomas. MYH is abundant in neurons where mitochondrial genomes exposed to reactive oxygen species (ROS) that damage DNA must maintain integrity over the entire mammalian life span.Specifications
| MUTYH | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q8R5G2, Q99P21, Q9UIF7 | |
| 170841, 4595, 70603 | |
| MUTYH Fusion Protein Ag7526 | |
| Primary | |
| -20°C | |
| MUTYH |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hMYH, MutY homolog, mutY homolog (E. coli), MUTYH, MYH | |
| MUTYH | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title