missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUSK Mouse anti-Mouse, Rat, Clone: 1429CT456.173.44, Abnova™
Mouse monoclonal antibody raised against mouse Musk.
Brand: Abnova MAB12362.400uL
This item is not returnable.
View return policy
Specifications
| MUSK | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:25) Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
| MGC117677/Mdk4/Mlk/Nsk1/Nsk2/Nsk3 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 18198 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1429CT456.173.44 | |
| Mouse monoclonal antibody raised against mouse Musk. | |
| In PBS (0.09% sodium azide) | |
| Musk | |
| Recombinant protein corresponding to mouse Musk. | |
| 400 μL | |
| Primary | |
| Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction