missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Antibody (S08-6B4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20013-100ul
This item is not returnable.
View return policy
Description
Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Muscle Phosphofructokinase/PFKM/PFK-1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| S08-6B4 | |
| Unconjugated | |
| 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase | |
| A synthetic peptide of human Muscle Phosphofructokinase/PFKM/PFK-1 (Uniprot # P08237) | |
| 100 μg | |
| Lipid and Metabolism | |
| 5213 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction