missing translation for 'onlineSavingsMsg'
Learn More

Muscarinic Acetylcholine Receptor M5/CHRM5 Antibody (590123), PE/Cy5.5, Novus Biologicals™

Product Code. 30038700 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30038700 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30038700 Supplier Novus Biologicals Supplier No. FAB10323PECY55

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Muscarinic Acetylcholine Receptor M5/CHRM5 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M5/CHRM5 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muscarinic Acetylcholine Receptor M5/CHRM5
Applications Flow Cytometry
Classification Monoclonal
Clone 590123
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias cholinergic receptor, muscarinic 5, HM5, MGC41838, muscarinic acetylcholine receptor M5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CHRM5, Met1-Pro532, Accession # P08912
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1133
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.