Learn More
Multi 13 elements solution - Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U for ICP 0.001mg/L, Fisher chemical™
Product Code. 12966183 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. 12966183
Brand: Fisher Chemical MS 003 500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Chemical Name or Material Thirteen Elements
Packaging Bottle
Analyte or Component Names Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U
Physical Form Liquid
Concentration 0.001 mg/L
Type Limit Detection Multi-component Verification Solution
Element Be, Na, Mg, K, Ca, Fe, Co, Rh, In, Ba, Ce, Pb, U
Solvent or Matrix 2% HNO3
Quantity 500 mL