missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC4 Antibody (S05-7I9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19704-100ul
This item is not returnable.
View return policy
Description
MUC4 Monoclonal antibody specifically detects MUC4 in Human samples. It is validated for Western Blot
Specifications
| MUC4 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S05-7I9 | |
| Unconjugated | |
| Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin | |
| Recombinant protein of human MUC4 (Uniprot # Q99102) | |
| 100 μg | |
| Cancer | |
| 4585 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction