missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTSS1 Antibody (M7P3A7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50518-0.025ml
This item is not returnable.
View return policy
Description
MTSS1 Monoclonal antibody specifically detects MTSS1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MTSS1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin | |
| FLJ44694, KIAA0429Missing in metastasis protein, metastasis suppressor 1, metastasis suppressor protein 1, Metastasis suppressor YGL-1, MIMA, MIMB, MIMDKFZp781P2223, MTSS1 | |
| Peptide Sequence - KPSIPEEHRQ (amino acids 679 - 688) | |
| 0.025 mL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 9788 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| M7P3A7 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction