missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€ - 470.00€
Specifications
| Antigen | MTMR6 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672471
|
Novus Biologicals
NBP2-94705-0.02ml |
0.02 mL |
204.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687721
|
Novus Biologicals
NBP2-94705-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTMR6 Polyclonal antibody specifically detects MTMR6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MTMR6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 9107 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.1.3, EC 3.1.3.-, EC 3.1.3.48, myotubularin related protein 6, myotubularin-related protein 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 382-621 of human MTMR6 (NP_004676.3). RCGQLDGDPKEVSPVFTQFLECVWHLTEQFPQAFEFSEAFLLQIHEHIHSCQFGNFLGNCQKEREELKLKEKTYSLWPFLLEDQKKYLNPLYSSESHRFTVLEPNTVSFNFKFWRNMYHQFDRTLHPRQSVFNIIMNMNEQNKQLEKDIKDLESKIKQRKNKQTDGILTKELLHSVHPESPNLKTSLCFKEQTLLPVNDALRTIEGSSPADNRYSEYAEEFSKSEPAVVSLEYGVARMTC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title