missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTF1 Mouse anti-Human, Clone: MTF1/2649, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | MTF1 |
|---|---|
| Clone | MTF1/2649 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
MTF1 Monoclonal specifically detects MTF1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| MTF1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| metal regulatory transcription factor 1, metal-regulatory transcription factor 1, metal-responsive transcription factor 1, MGC23036, MRE-binding transcription factor, MRE-binding transcription factor-1, MTF-1, Transcription factor MTF-1, zinc regulatory factor, ZRF | |
| Recombinant full-length human MTF1 protein | |
| Primary | |
| Store at 4°C. |
| MTF1/2649 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 4520 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title