missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTCH2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | MTCH2 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18010596
|
Proteintech
CL488-16888-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18028945
|
Proteintech
CL488-16888-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MTCH2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MTCH2. This antibody recognizes human antigen. The MTCH2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| MTCH2 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| HSPC032, MIMP, MTCH2 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 23788 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title