Learn More
MTA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14682-1-AP-150UL
 View more versions of this product
Product Code. 16813444
  • 410.00€ / 150µL
Description
Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.
Specifications
Specifications
MTA3
Polyclonal
Unconjugated
MTA3
KIAA1266, MTA3
Rabbit
Antigen Affinity Chromatography
RUO
100362346, 116871, 57504
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q924K8, Q9BTC8
MTA3
MTA3 Fusion Protein Ag6400
150 μL
Primary
Human, Mouse, Rat
Antibody