missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00€ - 481.00€
Specifications
| Antigen | MTA2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:50-1:200, Knockout Validated |
| Applications | Western Blot, ChIP Assay, Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682411
|
Novus Biologicals
NBP2-93800-0.02ml |
0.02 mL |
201.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655880
|
Novus Biologicals
NBP2-93800-0.1ml |
0.1 mL |
481.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTA2 Polyclonal antibody specifically detects MTA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay (ChIP), Gene Knock-OutSpecifications
| MTA2 | |
| Western Blot, ChIP Assay, Gene Knock-Out | |
| Unconjugated | |
| Rabbit | |
| Cancer, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS (pH 7.3), 50% glycerol | |
| 9219 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:50-1:200, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-180 of human MTA2 (NP_004730.2). DSNAREFEEESKQPGVSEQQRHQLKHRELFLSRQFESLPATHIRGKCSVTLLNETDILSQYLEKEDCFFYSLVFDPVQKTLLADQGEIRVGCKYQAEIPDRLVEGESDNRNQQKMEMKVWD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title