missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93571-0.1ml
This item is not returnable.
View return policy
Description
MT-ND3 Polyclonal antibody specifically detects MT-ND3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MT-ND3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Complex I ND3 Subunit, EC 1.6.5.3, Mitochondrially Encoded NADH Dehydrogenase 3, MTND3, NADH Dehydrogenase 3, NADH Dehydrogenase Subunit 3, NADH Dehydrogenase, Subunit 3 (Complex I), NADH3, NADH-Ubiquinone Oxidoreductase Chain 3, ND3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse MT-ND3 (NP_904335.1). MNLYTVIFINILLSLTLILVAFWLPQMNLYSEKANPYECGFDPTSSARLPFSMKFFLVAITFLLFDLEIALLLPLPWAIQTIKTSTMMIMAFILVTILSL | |
| 0.1 mL | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4537 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction