missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
224.00€ - 481.00€
Specifications
| Antigen | MSH6 |
|---|---|
| Applications | Western Blot, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18392656
|
Novus Biologicals
NBP3-03860-20UL |
20 μL |
224.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18310996
|
Novus Biologicals
NBP3-03860-100UL |
100 μL |
481.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSH6 Polyclonal specifically detects MSH6 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
| MSH6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein | |
| Recombinant protein of human MSH6. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, DNA Repair, Mismatch Repair | |
| PBS with 50% glycerol, pH7.3. | |
| 2956 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title