missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98819-50ul
This item is not returnable.
View return policy
Description
MSH5 Polyclonal antibody specifically detects MSH5 in Human samples. It is validated for Western Blot
Specifications
| MSH5 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| DKFZp434C1615, G7, hMSH5, MGC2939, mutS (E. coli) homolog 5, mutS homolog 5 (E. coli), mutS protein homolog 5, MUTSH5, NG23 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human MSH5. | |
| 50 μg | |
| DNA Repair, Mismatch Repair, modENCODE Antibodies | |
| 4439 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction