Description
MSH3 Monoclonal antibody specifically detects MSH3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | MSH3 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | CL14064 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | Divergent upstream protein, DNA mismatch repair protein Msh3, DUC1, DUG, DUPMGC163306, hMSH3, Mismatch repair protein 1, MRP1MGC163308, mutS (E. coli) homolog 3, mutS homolog 3 (E. coli) |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from P20585, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |