missing translation for 'onlineSavingsMsg'
Learn More

MRPS7 Monoclonal Antibody (OTI5C8), TrueMAB™, OriGene

Product Code. 17552723
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17552723 30 μL 30µL
17532723 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17552723 Supplier Origene Technologies Supplier No. TA803552S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPS7 Monoclonal antibody specifically detects MRPS7 in Human samples. It is validated for Western Blot

Specifications

Antigen MRPS7
Applications Western Blot
Classification Monoclonal
Clone OTI5C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene VWF
Gene Accession No. P04275
Gene Alias 28S ribosomal protein S7, mitochondrial; 30S ribosomal protein S7 homolog; bMRP27a; bMRP-27a; mitchondrial ribosomal protein S7; mitochondrial ribosomal protein S7; Mitochondrial small ribosomal subunit protein uS7m; MRP-S; Mrps7; MRP-S7; ribosomal protein, mitochondrial, S7; Rpms7; RP-S7; S7mt
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 764-1263 of human VWF produced in SF9 cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.