missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS31 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93779-0.02ml
This item is not returnable.
View return policy
Description
MRPS31 Polyclonal antibody specifically detects MRPS31 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| MRPS31 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| 28S ribosomal protein S31, mitochondrial, IMOGN38Imogen 38, mitochondrial ribosomal protein S31, MRP-S31, S31mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 66-395 of human MRPS31 (NP_005821.2). ICSKKDKQSVRTEETSKETSESQDSEKENTKKDLLGIIKGMKVELSTVNVRTTKPPKRRPLKSLEATLGRLRRATEYAPKKRIEPLSPELVAAASAVADSLPFDKQTTKSELLSQLQQHEEESRAQRDAKRPKISFSNIISDMKVARSATARVRSRPELRIQFDEGYDNYPGQEKTDDLKKRKNIFTGKRLNIFDMMAVTKEAPETDTSPSLWDVEFAKQLATVNEQPLQNGFEELIQWTKEGKLWEFPINNEAGFDDDGSEFHEHIFLEKHLESFPKQGPIRHFMELVTCGLSKNPYLSVKQKVEHIEWFRNYFNEKKDILKESNIQFN | |
| 0.02 mL | |
| Endocrinology, Immunology | |
| 10240 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction