missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35427-100ul
This item is not returnable.
View return policy
Description
MRPS2 Polyclonal antibody specifically detects MRPS2 in Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| MRPS2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| 28S ribosomal protein S2, mitochondrial, CGI-91, mitochondrial 28S ribosomal protein S2, mitochondrial ribosomal protein S2, MRP-S2, S2mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-296 of human MRPS2 (NP_057118.1).,, Sequence:, MATSSAALPRILGAGARAPSRWLGFLGKATPRPARPSRRTLGSATALMIRESEDSTDFNDKILNEPLKHSDFFNVKELFSVRSLFDARVHLGHKAGCRHRFMEPYIFGSRLDHDIIDLEQTATHLQLALNFTAHMAYRKGIILFISRNRQFSYLIENMARDCGEYAHTRYFRGGMLTNARLLFGPTVRLPDLIIFLHTLNNIFEPHVAVRDAAKMNIPTVGIVDTNCNPCLITYPVPGNDDSPLAVHLYCRLFQTAITRAKEKRQQVEALYRLQGQKEPGDQGPAHPPGADMSHSL | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51116 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction