missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL42 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35273-20ul
This item is not returnable.
View return policy
Description
MRPL42 Polyclonal antibody specifically detects MRPL42 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| MRPL42 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, ELISA | |
| 39S ribosomal protein L31, mitochondrial, 39S ribosomal protein L42, mitochondrial, HSPC204,28S ribosomal protein S32, mitochondrial, L31mt, mitochondrial ribosomal protein L42, mitochondrial ribosomal protein S32, MRPL31L42mt, MRP-L31MRP-S32, MRPS32MRP-L42, PTD007, RPML31S32mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-142 of human MRPL42 (NP_054769.1).,, Sequence:, MAVAAVKWVMSKRTILKHLFPVQNGALYCVCHKSTYSPLPDDYNCNVELALTSDGRTIVCYHPSVDIPYEHTKPIPRPDPVHNNEETHDQVLKTRLEEKVEHLEEGPMIEQLSKMFFTTKHRWYPHGRYHRCRKNLNPPKDR | |
| 20 μL | |
| Stem Cell Markers | |
| 28977 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction