missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP4/ABCC4 Antibody (ABCC4/9018), Novus Biologicals™
Mouse Monoclonal Antibody
245.00€ - 462.00€
Specifications
| Antigen | MRP4/ABCC4 |
|---|---|
| Clone | ABCC4/9018 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627544
|
Novus Biologicals
NBP3-23892-20ug |
20 μg |
245.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651015
|
Novus Biologicals
NBP3-23892-100ug |
100 μg |
462.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRP4/ABCC4 Monoclonal antibody specifically detects MRP4/ABCC4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MRP4/ABCC4 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 10257 | |
| IgG2b κ | |
| Protein A or G purified |
| ABCC4/9018 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATP-binding cassette sub-family C member 4, ATP-binding cassette, sub-family C (CFTR/MRP), member 4, EST170205, MOAT-BbA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4), MOATBcanalicular multispecific organic anion transporter (ABC superfamily), MRP4MRP/cMOAT-related ABC transporter, multidrug resistance-associated protein 4, multispecific organic anion transporter B, Multi-specific organic anion transporter B | |
| Recombinant fragment (around aa1-200) of human MRP4/ABCC4 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title