Learn More
MRP1 ABCC1 Mouse anti-Human, Unconjugated, Clone: 1G4A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67228-1-IG-20UL
Description
The MRP1, ABCC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MRP1, ABCC1. This antibody recognizes human antigen. The MRP1, ABCC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MRP1 ABCC1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot | |
| 1G4A2 | |
| Unconjugated | |
| 20 μL | |
| 4363 | |
| IgG1 |