Learn More
MRP1 ABCC1 Mouse anti-Human, Unconjugated, Clone: 1G4A2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67228-1-IG-20UL
 View more versions of this product
Product Code. 16849755
  • 145.00€ / 20µL
Description
The MRP1, ABCC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MRP1, ABCC1. This antibody recognizes human antigen. The MRP1, ABCC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MRP1 ABCC1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot
1G4A2
Unconjugated
20 μL
4363