missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRAS Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148733
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.
Specifications
| MRAS | |
| Polyclonal | |
| Unconjugated | |
| MRAS | |
| 2900078C09Rik; AI326250; MRAS; M-RAs; Muscle and microspikes Ras; muscle RAS oncogene homolog; ras-related protein M-Ras; Ras-related protein R-Ras3; RRAS3; R-RAS3; Xras; X-Ras | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 22808, 25482 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O14807, P97538 | |
| MRAS | |
| E. coli-derived recombinant human M-Ras, Met1-Leu208 (Lys181Glu). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction