Learn More
Heidolph™ MR Hei-Mix S Magnetic Stirrer
Product Code. 15117004 Shop All Heidolph Instruments Products
Change view
Click to view available options
Dimensions (L x W x H):
140 x 126 x 80 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
15117004 140 x 126 x 80 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15117004 Supplier Heidolph™ Supplier No. 5030200000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Stir quickly and efficiently with an extended speed range magnetic stirrer. Heidolph™ MR Hei-Mix S Magnetic Stirrer is a space-saving, unheated unit.
  • For gentle stirring in biology and microbiology applications.
  • Stir quickly and efficiently with an extended speed range.
  • Space-saving unit for efficiently mixing samples.
  • Small footprint.
  • Ideal for titrations due to white-colored top plate.
  • Long-lasting housing and top plate material.
TRUSTED_SUSTAINABILITY
Specifications
Speed 2200 rpm
Dimensions (L x W x H) 140 x 126 x 80 mm
Top Plate Diameter (Metric) 104 mm
Description Unheated stir plate
Type Magnetic Stirrer