Learn More
MPZL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16839093 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16839093 20 μL 20µL
16829093 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16839093 Supplier Proteintech Supplier No. 117871AP20UL
Rabbit Polyclonal Antibody
This antibody can be used to detect endogenous MPZL2, however, the reason for the additional band of 32 kDa is still unknown.
Epithelial V-like antigen (EVA) is expressed in thymus epithelium and strongly downregulated by thymocyte developmental progression. This gene is expressed in the thymus and in several epithelial structures early in embryogenesis.
Specifications
Antigen MPZL2
Applications Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MPZL2
Gene Accession No. O60487, O70255
Gene Alias Epithelial V like antigen 1, EVA, EVA1, MPZL2, myelin protein zero like 2
Gene Symbols MPZL2
Host Species Rabbit
Immunogen MPZL2 Fusion Protein Ag2419
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10205, 14012, 300679
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG