missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPRIP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94741-0.1ml
This item is not returnable.
View return policy
Description
MPRIP Polyclonal antibody specifically detects MPRIP in Human samples. It is validated for Western Blot
Specifications
| MPRIP | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| KIAA0864Rho interacting protein 3, MRIPP116RIP, myosin phosphatase Rho interacting protein, p116Ripmyosin phosphatase-Rho interacting protein, RHOIP3myosin phosphatase Rho-interacting protein, RIP3M-RIPRho-interacting protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human MPRIP (NP_055949.2). MSAAKENPCRKFQANIFNKSKCQNCFKPRESHLLNDEDLTQAKPIYGGWLLLAPDGTDFDNPVHRSRKWQRRFFILYEHGLLRYALDEMPTTLPQGTINMNQCTDVVDGEGRTGQKFSLCILTPEKEHFIRAETKEIVSGWLEMLMVYPRTNKQNQKKKRKVEPPTPQEPGPAKVAVTSSSSSSSSSSSIPSAEKVPTTKSTLWQEEMRTKDQPDGSSLSPAQSPSQSQPPAASSLREPGLESKEEESAMSSDRMDCGRKVRVESGYFSLEKTKQDLKAEEQQLPPPLSPPSPSTPNHRRSQVIEKFEAL | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 23164 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto