missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | MPP2 |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233377
|
Proteintech
13589-1-AP-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237166
|
Proteintech
13589-1-AP-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MPP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MPP2. This antibody recognizes human, mouse, rat antigen. The MPP2 antibody has been validated for the following applications: WB, IP, IHC, ELISA analysis.Specifications
| MPP2 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| DKFZp686A06252; DKFZp686J2189; DKFZp761D0712; DLG2 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 600 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 4355 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title