Learn More
MP Biomedicals™ L-Broth Agar, Capsules
Product Code. 11363209 Shop All MP Biomedical Products
Click to view available options
Quantity:
1 kg
454 g
Unit Size:
1kg
454g
Product Code. 11363209
Brand: MP Biomedicals™ 113001221
For the growth of E.coli and certified for plasmid and other bacterial cultures in liquid medium
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 15° to 30°C
Product Type Agar
Sterility Sterile
pH 6.5
Quantity 454 g
compliance-icons
Product Identifier
  • L-Broth Agar Capsules
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : agar