Learn More
MP Biomedicals™ Immunidiffusion (RID) Plates
Product Code. 11403626 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
12 Pk.
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403626 12 Pk. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11403626 Supplier MP Biomedicals™ Supplier No. 08421651
  • ONLINE EXCLUSIVE PRICE
    111.00€
    105.00€ / Pack of 12
    Save 6.00€
    5% Off
Estimated Shipment: 10-02-2027
Used for double and single radial immunodiffusion techniques
  • Polystyrene construction with close-fitting covers to minimize gel dehydration
  • Comes with recessed bottoms to prevent scratching on bench surfaces
  • Hydrophilic in nature for easy gel pouring and better adherence
Tekniske data
Material Polystyrene
Type Immunodiffusion Plate
For Use With (Application) Double and single radial immunodiffusion techniques
Quantity 12 Pk.
Storage Requirements 15° to 30°C
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.