missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Complement C1q Protein
Shop All MP Biomedical Products
Click to view available options
:
1mL
Description
Purity: Functionally and biochemically pure by PAGE. Concentration: 1mg/mL. Supplied in PBS, 40% glycerol.
Specifications
Specifications
| Concentration | 1.1mg/mL |
| Formulation | Supplied in PBS, 40% Glycerol |
| Name | Human Complement C1q |
| Purification Method | SDS-PAGE |
| Quantity | 1mL |
| Source | Human Serum |
| Storage Requirements | -70°C |
| Cross Reactivity | Human Serum |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido