missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CellMaxx™ Bovine Albumin, Low IgG, Stem Cell Culture

Product Code. 17531096 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1000 g
Unit Size:
100g
10g
1kg
25g
500g
50g
Product Code. Quantity unitSize
17531096 10 g 10g
17541096 25 g 25g
17551096 50 g 50g
11431655 100 g 100g
17561096 500 g 500g
17571096 1000 g 1kg
6 options
This item is not returnable. View return policy

Product Code. 17531096

Brand: MP Biomedicals™ 0219989710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation

Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells; IgG removed to very low levels. Lipid-rich formulation can be substituted for Albumax I™.

Physical characteristics:

  • Form: Off white to brown powder
  • Purity: ≥98% (w/w)
  • Total Protein: ≥96%
  • IgG 0.005% w/w
  • pH (4%): 6.5 to 7.5
  • Solubility: Dissolves in 20 minutes at 20° to 25°C
  • Moisture: ≤5% w/w
Microbial evaluation and control:
  • Endotoxin: ≤1EU/mg
  • Mycoplasma: Negative
  • Bioburden: ≤100cfu/g
  • Virus (9CFR): Negative
  • Reported: Lipid content
Certifications: ISO9000, cGMP, EDQM

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Mammalian Cell Culture, Western Blot
Formulation Lipid-rich
Molecular Weight (g/mol) 66
Name CellMaxx™ Bovine Albumin, Low IgG
pH Range 6.5 to 7.5
Purification Method Chromatography
Quantity 10 g
Source Bovine
Storage Requirements 4°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.