missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Aprotinin, From Bovine Lung

Product Code. 11426171 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426171 10 mg 10mg
11436171 25 mg 25mg
11446171 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426171 Supplier MP Biomedicals™ Supplier No. 0219115810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From Bovine Lung

Aprotinin, [9087-70-1]

  • C284H432N84O79S7
  • Lyophilized white powder

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Inhibition Assay
Molecular Weight (g/mol) 6511.23
Name Aprotinin from Bovine Lung
pH Range 5 to 7
Quantity 10 mg
Source Bovine Lung
Storage Requirements 4°C
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.