missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-92134-25ul
This item is not returnable.
View return policy
Description
MOX1 Polyclonal specifically detects MOX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MOX1 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| P50221 | |
| MEOX1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:WGCLRNPHSEGNGASGLPHYPPTPFSFHQKPDFLATATAAYPDFSASCLAATPHSLPQEEHIFTEQHPAFPQSPNWHFPVSDARRRPNSGPAGGSKEMGTSSLGLVDTTGGPGDDYGVLGSTANETEKKSSR | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| homeobox protein MOX-1, mesenchyme homeobox 1MOX1mesenchyme homeo box 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4222 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction