Learn More
Mouse YM1/Chitinase 3-like 3 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 15 publications
Brand:  R&D Systems AF2446-SP
Product Code. 18706947
  • 241.00€ / 25µg
Description
YM1/Chitinase 3-like 3 Polyclonal specifically detects YM1/Chitinase 3-like 3 in Mouse samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
YM1/Chitinase 3-like 3
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AI505981, Chi3l3, Chitinase 3 like 3, Chitinase 3-like 3, ECF-L, Ym1
Goat
Affinity Purified
RUO
1117
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.2 ug/mL, Simple Western 2.5 ug/mL
O35744
CHI3L2
Mouse myeloma cell line NS0-derived recombinant mouse YM1/Chitinase 3-like 3 Tyr22-Tyr398 Accession # O35744
25 μg
Primary
Detects mouse YM1/Chitinase 3-like 3 in direct ELISAs and Western blots.