missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VEGF164 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 79 publications
Brand: R&D Systems AF-493-NA
This item is not returnable.
View return policy
Description
VEGF Polyclonal specifically detects VEGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| VEGF | |
| Polyclonal | |
| Unconjugated | |
| AAA40547 | |
| VEGFA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse VEGF 164 Ala27-Arg190 Accession # AAA40547 | |
| 100 μg | |
| Primary | |
| Detects mouse VEGF in ELISAs and Western blots. In Western blots, detection of recombinant mouse (rm) VEGF 164 rmVEGF 120 rmVEGF 188 and recombinant rat VEGF 164 is observered but no cross-reactivity with rmVEGF-B 167, rmVEGF-B 186 or recombinant human (rh) VEGF 165 is observed. In sandwich immunoassays, less than 0.04% cross-reactivity with rhVEGF 121 rhVEGF 165 and rhVEGF/PlGF is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.05-0.15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7422 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction