missing translation for 'onlineSavingsMsg'
Learn More

Mouse TWEAK/TNFSF12 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18715737
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18715737 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18715737 Supplier R&D Systems Supplier No. AF1237SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

TWEAK/TNFSF12 Polyclonal specifically detects TWEAK/TNFSF12 in Mouse samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWEAK/TNFSF12
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 4-16 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O54907
Gene Alias APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, TNFSF12, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12
Gene Symbols Tnfsf12
Host Species Goat
Immunogen E. coli-derived recombinant mouse TWEAK/TNFSF12 Arg105-His249 Accession # O54907
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8742
Test Specificity Detects TWEAK/TNFSF12 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human TWEAK is observed and less than 1% cross-reactivity with recombinant mouse (rm) BAFF, rmFas Ligand, rmOX40 Ligand, rmTNF-alpha , and rmTRAIL is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.