missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Tryptase epsilon/BSSP-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2059-SP
This item is not returnable.
View return policy
Description
Tryptase epsilon/BSSP-4/PRSS22 Monoclonal specifically detects Tryptase epsilon/BSSP-4/PRSS22 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Tryptase epsilon/BSSP-4/PRSS22 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BSP-2, BSSP4, BSSP-4, hBSSP-4, MGC9599, prosemin, protease, serine S1 family member 22, protease, serine, 22, PRSS22, serine protease 22, serine protease 26, SP001LA, tryptase epsilon, UNQ302/PRO343 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64063 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunoprecipitation | |
| 246744 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q7TML0 | |
| PRSS22 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tryptase epsilon/BSSP-4 Ala33-Ser306 (predicted) Accession # Q7TML0 | |
| 25 μg | |
| Primary | |
| Detects mouse Tryptase epsilon/BSSP-4 in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction