missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TREML2/TLT-2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6354
This item is not returnable.
View return policy
Description
TREML2/TLT-2 Polyclonal specifically detects TREML2/TLT-2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| TREML2/TLT-2 | |
| Polyclonal | |
| Unconjugated | |
| Q2LA85 | |
| TREML2 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TREML2/TLT-2 His25-Ala270 Accession # Q2LA85 | |
| 100 μg | |
| Primary | |
| Detects mouse TREML2/TLT-2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) TREML1, rmTREML4, rmTREM1, rmTREM2b, rmTREM3, rmTREM4, and recombinant human TREM2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| C6orf76, dJ238O23.1, FLJ13693, MGC149715, MGC149716, TLT2, TLT-2, trem-like transcript 2 protein, triggering receptor expressed on myeloid cells-like 2, triggering receptor expressed on myeloid cells-like protein 2, UNQ6268/PRO20473 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 79865 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction