missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TREM-1 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1187A
This item is not returnable.
View return policy
Description
TREM1 Monoclonal specifically detects TREM1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| TREM1 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54210 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 174031 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q9JKE2 | |
| TREM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREM-1 Ala21-Ser202 Accession # Q9JKE2 | |
| 100 Tests | |
| Primary | |
| Detects mouse TREM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse TREM-2B, recombinant human (rh) TREM-1, or rhTREM-2 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction