missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TIM-1/KIM-1/HAVCR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 22 publications
Brand: R&D Systems AF1817
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Polyclonal specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Western Blot.
Specifications
| TIM-1/KIM-1/HAVCR | |
| Polyclonal | |
| Unconjugated | |
| NP_001160104 | |
| HAVCR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIM-1/KIM-1/HAVCR Tyr22-Thr212 Accession # NP_001160104 | |
| 100 μg | |
| Primary | |
| Detects mouse TIM-1/KIM-1/HAVCR in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human TIM-1, recombinant mouse (rm) TIM-2, rmTIM-3, rmTIM-4, rmTIM-6, and rmTIM-7 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 26762 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction