missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TGN38 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB7944
This item is not returnable.
View return policy
Description
TGN38 Monoclonal specifically detects TGN38 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| TGN38 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q62313 | |
| TGOLN2 | |
| E. coli-derived recombinant mouse TGN38 His320-Leu349 Accession # Q62313 | |
| 100 μg | |
| Primary | |
| Detects mouse TGN38 in ELISAs. | |
| Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 831629 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TGN46, TGN46TGN38 homolog, TGN48MGC14722, TGN51, TGN5148, 51kD isoforms), Tgoln1, Tgoln2, trans-golgi network protein 2, Trans-Golgi network protein TGN51, Ttgn1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10618 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction