missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IL-15
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
- Interleukin 15 (IL-15) is a four alpha-helix bundle cytokine with many similar properties to IL-2, with which it shares components of its receptor
- IL-15 is produced by antigen-presenting cells, T cells, and B cells, as well as epithelial and stromal cells
- The IL-15 receptor is a heterotrimeric receptor composed of IL-15Ra (the high-affinity receptor for IL-15), as well as IL-2/IL-15Rb (CD122) and common gamma chain (CD132)
- IL-15 binds to IL-15Ra and can then be presented in trans to IL-2/IL-15Rb and common gamma chain on other cells
- Trans-presentation is thought to be the major mechanism by which IL-15-mediated responses occur in mice, although may not be necessary in humans
- The cytoplasmic domains of IL-2/IL-15Rb and common gamma chain mediate signaling to activate JAK/STAT and PI3K pathways
- IL-15 supports the survival and proliferation of CD8+ T cells, and promotes homeostasis of memory T cells
Specifications
Specifications
| Name | IL-T, Interleukin-15 |
| Quantity | 100 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; Lymphocytes; NK Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction