missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat Themis Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6816
This item is not returnable.
View return policy
Description
Themis Polyclonal specifically detects Themis in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Themis | |
| Polyclonal | |
| Unconjugated | |
| Q8BGW0 | |
| THEMIS | |
| E. coli-derived recombinant mouse Themis Lys122-Lys237 Accession # Q8BGW0 | |
| 100 μg | |
| Primary | |
| Detects mouse Themis in direct ELISAs and mouse and rat Themis in Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human Themis is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, SPOT, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 387357 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction