missing translation for 'onlineSavingsMsg'
Learn More

Mouse/Rat Kallikrein 7 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18769197
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18769197 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18769197 Supplier R&D Systems Supplier No. AF7688SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Kallikrein 7 Polyclonal specifically detects Kallikrein 7 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 7
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q91VE3
Gene Alias EC 3.4.21, EC 3.4.21.117, hK7, hSCCE, kallikrein 7 (chymotryptic, stratum corneum), kallikrein-related peptidase 7, KLK7, protease, serine, 6, PRSS6, SCCEkallikrein-7, Serine protease 6, signal protein, Stratum corneum chymotryptic enzyme
Gene Symbols KLK7
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Kallikrein 7 Gln22-Arg249 (predicted) Accession # Q91VE3
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5650
Test Specificity Detects mouse and rat Kallikrein 7 in Western blots. Detects mouse Kallikrein 7 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Kallikrein 7 is observed, and less than 1% cross-reactivity with recombinant mouse Kallikrein 5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.