missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PILR-alpha Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4318-SP
This item is not returnable.
View return policy
Description
PILR-alpha Polyclonal specifically detects PILR-alpha in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| PILR-alpha | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29992 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q2YFS3 | |
| PILRA | |
| Mouse myeloma cell line NS0-derived recombinant mouse PILR-alpha isoform 1 (R&D Systems, Catalog # 4318-PR) Leu21-Val197 Accession # Q2YFS3 | |
| 25 μg | |
| Primary | |
| Detects mouse PILR-alpha in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse PILR-beta is observed and 5% cross-reactivity with recombinant human PILR-alpha is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering