missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PD-L1/B7-H1 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
247.00€ - 595.00€
Specifications
| Antigen | PD-L1 |
|---|---|
| Clone | 2096A |
| Dilution | Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 5 μg/mL |
| Applications | Immunohistochemistry, Multiplex Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306087
|
R&D Systems
MAB11630-100 |
100 μg |
595.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306072
|
R&D Systems
MAB11630-SP |
25 μg |
247.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Mouse samples. It is validated for Immunohistochemistry,Multiplex ImmunofluorescenceSpecifications
| PD-L1 | |
| Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Mouse myeloma cell line, NS0 derived mouse PD-L1/B7-H1, Phe19-His239, Accession # Q9EP73 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2096A | |
| Immunohistochemistry, Multiplex Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q9EP73 | |
| 29126 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title