missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NKp46/NCR1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 29 publications
Brand: R&D Systems AF2225
This item is not returnable.
View return policy
Description
NKp46/NCR1 Polyclonal specifically detects NKp46/NCR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, CyTOF-ready.
Specifications
| NKp46/NCR1 | |
| Polyclonal | |
| Unconjugated | |
| Q8C567 | |
| NCR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse NKp46/NCR1 Glu22-Asn255 Accession # Q8C567 | |
| 100 μg | |
| Agonist Activity | |
| 9437 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, Agonist Activity 0.4-2.4 ug/mL, CyTOF-ready | |
| CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects mouse NKp46/NCR1 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) NKp46 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction